Anti-WDR18 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25496EA 518 EUR
ABNOPAB25496
Anti-WDR18 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of WDR18.

Recommended Dilutions: Western Blot: 0.5-1 ?g/ml; Immunohistochemistry: 2.5 ?g/ml

Type: Primary
Antigen: WDR18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR