Anti-DBX2 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25483EA 454 EUR
ABNOPAB25483
Anti-DBX2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of DBX2.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry: 5 ?g/ml

Type: Primary
Antigen: DBX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR