Anti-ABCD2 Goat Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25454EA 518 EUR
ABNOPAB25454
Anti-ABCD2 Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody raised against synthetic peptide of ABCD2.

Recommended Dilutions: ELISA: 1:32000; Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: ABCD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR