Anti-ADAR Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB18055EA 419 EUR
ABNOPAB18055
Anti-ADAR Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of ADAR.

Recommended Dilutions: Immunohistochemistry: 1:50~1:100 IELISA: 1:40000

Type: Primary
Antigen: ADAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR