Anti-MAP2K2 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25229EA 370 EUR
ABNOPAB25229
Anti-MAP2K2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAP2K2.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100; Western Blot: 1:500-1:1000; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: MAP2K2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR