Anti-IL16 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25188EA 454 EUR
ABNOPAB25188
Anti-IL16 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of IL16.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: IL16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR