Anti-DEFA1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25168EA 518 EUR
ABNOPAB25168
Anti-DEFA1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of DEFA1.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: DEFA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR