Anti-MAPK3 Mouse Monoclonal Antibody [clone: 1E5]

Lieferant: Abnova
ABNOMAB10800EA 454 EUR
ABNOMAB10800
Anti-MAPK3 Mouse Monoclonal Antibody [clone: 1E5]
Antikörper
Mouse monoclonal antibody raised against partial recombinant MAPK3.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400

Type: Primary
Antigen: MAPK3
Clonality: Monoclonal
Clone: 1E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR