Anti-LCN1 Mouse Monoclonal Antibody [clone: 10B10]

Lieferant: Abnova
ABNOMAB10788EA 370 EUR
ABNOMAB10788
Anti-LCN1 Mouse Monoclonal Antibody [clone: 10B10]
Antikörper
Mouse monoclonal antibody raised against partial recombinant LCN1.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000

Type: Primary
Antigen: LCN1
Clonality: Monoclonal
Clone: 10B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR