Anti-ITGAE Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25149EA 454 EUR
ABNOPAB25149
Anti-ITGAE Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic peptide of ITGAE.

Recommended Dilutions: Western Blot: 1:500-1:1008)

Type: Primary
Antigen: ITGAE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR