Anti-INHA Mouse Monoclonal Antibody [clone: 4E2]

Lieferant: Abnova
ABNOMAB10700EA 370 EUR
ABNOMAB10700
Anti-INHA Mouse Monoclonal Antibody [clone: 4E2]
Antikörper
Mouse monoclonal antibody raised against partial recombinant INHA.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunofluorescence: 1:200-1:1000

Type: Primary
Antigen: INHA
Clonality: Monoclonal
Clone: 4E2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR