Anti-IGF1R Mouse Monoclonal Antibody [clone: 3G5C1]

Lieferant: Abnova
ABNOMAB10693EA 370 EUR
ABNOMAB10693
Anti-IGF1R Mouse Monoclonal Antibody [clone: 3G5C1]
Antikörper
Mouse monoclonal antibody raised against partial recombinant IGF1R.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000

Type: Primary
Antigen: IGF1R
Clonality: Monoclonal
Clone: 3G5C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR