Anti-MAP3K5 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25068EA 568 EUR
ABNOPAB25068
Anti-MAP3K5 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAP3K5.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: MAP3K5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR