Anti-EIF2AK2 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25056EA 568 EUR
ABNOPAB25056
Anti-EIF2AK2 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic phosphopeptide of EIF2AK2.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: EIF2AK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR