Anti-GSK3B Mouse Monoclonal Antibody [clone: 3D10]
Lieferant: Abnova
ABNOMAB10672EA 518 EUR
ABNOMAB10672
Anti-GSK3B Mouse Monoclonal Antibody [clone: 3D10]
Antikörper
Mouse monoclonal antibody raised against partial recombinant GSK3B.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400
Type: Primary
Antigen: GSK3B
Clonality: Monoclonal
Clone: 3D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey, Mouse, Rat