Anti-GOT2 Mouse Monoclonal Antibody [clone: 3E9]

Lieferant: Abnova
ABNOMAB10669EA 370 EUR
ABNOMAB10669
Anti-GOT2 Mouse Monoclonal Antibody [clone: 3E9]
Antikörper
Mouse monoclonal antibody raised against partial recombinant GOT2.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunofluorescence: 1:200-1:1000

Type: Primary
Antigen: GOT2
Clonality: Monoclonal
Clone: 3E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR