Anti-FGFR4 Mouse Monoclonal Antibody [clone: 4H2B10B2]

Lieferant: Abnova
ABNOMAB10648EA 518 EUR
ABNOMAB10648
Anti-FGFR4 Mouse Monoclonal Antibody [clone: 4H2B10B2]
Antikörper
Mouse monoclonal antibody raised against partial recombinant FGFR4.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000

Type: Primary
Antigen: FGFR4
Clonality: Monoclonal
Clone: 4H2B10B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR