Anti-FCER2 Mouse Monoclonal Antibody [clone: 5B5]
Lieferant: Abnova
ABNOMAB10644EA 518 EUR
ABNOMAB10644
Anti-FCER2 Mouse Monoclonal Antibody [clone: 5B5]
Antikörper
Mouse monoclonal antibody raised against partial recombinant FCER2.
Recommended Dilutions: ELISA: 1:10000; Flow Cytometry: 1:200-1:400
Type: Primary
Antigen: FCER2
Clonality: Monoclonal
Clone: 5B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human