Anti-FBLN5 Mouse Monoclonal Antibody [clone: 3F8D5,7F8D5]

Lieferant: Abnova
ABNOMAB10643EA 350 EUR
ABNOMAB10643
Anti-FBLN5 Mouse Monoclonal Antibody [clone: 3F8D5,7F8D5]
Antikörper
Mouse monoclonal antibody raised against partial recombinant FBLN5.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: FBLN5
Clonality: Monoclonal
Clone: 3F8D5,7F8D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR