Anti-BRCA1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25020EA 454 EUR
ABNOPAB25020
Anti-BRCA1 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic peptide of BRCA1.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: BRCA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR