Anti-RAD51 Rat Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24992EA 518 EUR
ABNOPAB24992
Anti-RAD51 Rat Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of RAD51.

Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:200; Immunofluorescence: 1:50-1:200

Type: Primary
Antigen: RAD51
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR