Anti-CYP2D6 Rat Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24990EA 518 EUR
ABNOPAB24990
Anti-CYP2D6 Rat Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of CYP2D6.

Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: CYP2D6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR