Anti-DKK1 Mouse Monoclonal Antibody [clone: 2H2]

Lieferant: Abnova
ABNOMAB10624EA 419 EUR
ABNOMAB10624
Anti-DKK1 Mouse Monoclonal Antibody [clone: 2H2]
Antikörper
Mouse monoclonal antibody raised against partial recombinant DKK1.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: DKK1
Clonality: Monoclonal
Clone: 2H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR