Anti-CRTC1 Mouse Monoclonal Antibody [clone: 1B5]

Lieferant: Abnova
ABNOMAB10614EA 370 EUR
ABNOMAB10614
Anti-CRTC1 Mouse Monoclonal Antibody [clone: 1B5]
Antikörper
Mouse monoclonal antibody raised against partial recombinant CRTC1.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400

Type: Primary
Antigen: CRTC1
Clonality: Monoclonal
Clone: 1B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR