Anti-TNFRSF6B Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24965EA 454 EUR
ABNOPAB24965
Anti-TNFRSF6B Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of TNFRSF6B.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Immunofluorescence: 1:50-1:200

Type: Primary
Antigen: TNFRSF6B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR