Anti-MPO Mouse Monoclonal Antibody [clone: 9B12G7,4D8B12,9B12D9,9C11A5]
Lieferant: Abnova
ABNOMAB10590EA 518 EUR
ABNOMAB10590
Anti-MPO Mouse Monoclonal Antibody [clone: 9B12G7,4D8B12,9B12D9,9C11A5]
Antikörper
Mouse monoclonal antibody raised against partial recombinant MPO.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000
Type: Primary
Antigen: MPO
Clonality: Monoclonal
Clone: 9B12G7,4D8B12,9B12D9,9C11A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human