Anti-CRK Mouse Monoclonal Antibody [clone: 3G11C1]

Lieferant: Abnova
ABNOMAB10572EA 454 EUR
ABNOMAB10572
Anti-CRK Mouse Monoclonal Antibody [clone: 3G11C1]
Antikörper
Mouse monoclonal antibody raised against partial recombinant CRK.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400

Type: Primary
Antigen: CRK
Clonality: Monoclonal
Clone: 3G11C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR