Anti-iHog Mouse Monoclonal Antibody [clone: 3G8]
Lieferant: Abnova
ABNOMAB10568EA 518 EUR
ABNOMAB10568
Anti-iHog Mouse Monoclonal Antibody [clone: 3G8]
Antikörper
Mouse monoclonal antibody raised against partial recombinant iHog.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000
Type: Primary
Antigen: iHog
Clonality: Monoclonal
Clone: 3G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Drosophila