Anti-FLT1 Mouse Monoclonal Antibody [clone: 3D10]

Lieferant: Abnova
ABNOMAB10557EA 370 EUR
ABNOMAB10557
Anti-FLT1 Mouse Monoclonal Antibody [clone: 3D10]
Antikörper
Mouse monoclonal antibody raised against partial recombinant FLT1.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: FLT1
Clonality: Monoclonal
Clone: 3D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR