Anti-CHD3 Mouse Monoclonal Antibody [clone: 2G4]
Lieferant: Abnova
ABNOMAB10553EA 518 EUR
ABNOMAB10553
Anti-CHD3 Mouse Monoclonal Antibody [clone: 2G4]
Antikörper
Mouse monoclonal antibody raised against partial recombinant CHD3.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400
Type: Primary
Antigen: CHD3
Clonality: Monoclonal
Clone: 2G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse