Anti-UCHL1 Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24946EA 772 EUR
ABNOPAB24946
Anti-UCHL1 Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of PGP 9.5.

Recommended Dilutions: Western Blot: 1:1000; Immunohistochemistry: 1:5000

Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR