Anti-CRH Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24940EA 518 EUR
ABNOPAB24940
Anti-CRH Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of CRH.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: CRH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR