Anti-CD40 Mouse Monoclonal Antibody [clone: 9G10]

Lieferant: Abnova
ABNOMAB10537EA 370 EUR
ABNOMAB10537
Anti-CD40 Mouse Monoclonal Antibody [clone: 9G10]
Antikörper
Mouse monoclonal antibody raised against partial recombinant CD40.

Recommended Dilutions: ELISA: 1:10000; Immunofluorescence: 1:200-1:1000

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: 9G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR