Anti-CD276 Mouse Monoclonal Antibody [clone: 6A1]

Lieferant: Abnova
ABNOMAB10532EA 370 EUR
ABNOMAB10532
Anti-CD276 Mouse Monoclonal Antibody [clone: 6A1]
Antikörper
Mouse monoclonal antibody raised against partial recombinant CD276.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400

Type: Primary
Antigen: CD276
Clonality: Monoclonal
Clone: 6A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR