Anti-CCT2 Mouse Monoclonal Antibody [clone: 5B5C4]

Lieferant: Abnova
ABNOMAB10523EA 454 EUR
ABNOMAB10523
Anti-CCT2 Mouse Monoclonal Antibody [clone: 5B5C4]
Antikörper
Mouse monoclonal antibody raised against partial recombinant CCT2.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400

Type: Primary
Antigen: CCT2
Clonality: Monoclonal
Clone: 5B5C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR