Anti-CASP8 Mouse Monoclonal Antibody [clone: 1H11]

Lieferant: Abnova
ABNOMAB10519EA 454 EUR
ABNOMAB10519
Anti-CASP8 Mouse Monoclonal Antibody [clone: 1H11]
Antikörper
Mouse monoclonal antibody raised against partial recombinant CASP8.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Flow Cytometry: 1:200-1:400

Type: Primary
Antigen: CASP8
Clonality: Monoclonal
Clone: 1H11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR