Anti-NPPB Mouse Monoclonal Antibody [clone: 8D5B4C11]

Lieferant: Abnova
ABNOMAB10509EA 454 EUR
ABNOMAB10509
Anti-NPPB Mouse Monoclonal Antibody [clone: 8D5B4C11]
Antikörper
Mouse monoclonal antibody raised against synthetic peptide of NPPB.

Recommended Dilutions: ELISA: 1:10000; Immunohistochemistry: 1:200-1:1000

Type: Primary
Antigen: NPPB
Clonality: Monoclonal
Clone: 8D5B4C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR