Anti-BMX Mouse Monoclonal Antibody [clone: 1C6]

Lieferant: Abnova
ABNOMAB10508EA 370 EUR
ABNOMAB10508
Anti-BMX Mouse Monoclonal Antibody [clone: 1C6]
Antikörper
Mouse monoclonal antibody raised against partial recombinant BMX.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: BMX
Clonality: Monoclonal
Clone: 1C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR