Anti-BMP4 Mouse Monoclonal Antibody [clone: 10F4B4]

Lieferant: Abnova
ABNOMAB10507EA 518 EUR
ABNOMAB10507
Anti-BMP4 Mouse Monoclonal Antibody [clone: 10F4B4]
Antikörper
Mouse monoclonal antibody raised against partial recombinant BMP4.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: BMP4
Clonality: Monoclonal
Clone: 10F4B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR