Anti-TFRC Mouse Monoclonal Antibody [clone: CY-TFR]

Lieferant: Abnova
ABNOMAB6780EA 454 EUR
ABNOMAB6780
Anti-TFRC Mouse Monoclonal Antibody [clone: CY-TFR]
Antikörper
Mouse monoclonal antibody raised against TFRC/TFR2.

Type: Primary
Antigen: TFRC
Clonality: Monoclonal
Clone: CY-TFR
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR