Anti-Inactivated EV71 Mouse Monoclonal Antibody [clone: 3F4]

Lieferant: Abnova
ABNOMAB5639-M07EA 382 EUR
ABNOMAB5639-M07
Anti-Inactivated EV71 Mouse Monoclonal Antibody [clone: 3F4]
Antikörper
Mouse monoclonal antibody raised against inactivated EV71.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: Inactivated EV71
Clonality: Monoclonal
Clone: 3F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR