Anti-ABCC1 Mouse Monoclonal Antibody [clone: QCRL-4]

Lieferant: Abnova
ABNOMAB5158EA 1030 EUR
ABNOMAB5158
Anti-ABCC1 Mouse Monoclonal Antibody [clone: QCRL-4]
Antikörper
Mouse monoclonal antibody raised against native ABCC1.

Recommended Dilutions: Flow Cytometry: 1:20-1:40; Dot Blot: 1:10000

Type: Primary
Antigen: ABCC1
Clonality: Monoclonal
Clone: QCRL-4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR