Anti-ABCC1 Monoclonal Antibody [clone: QCRL-3]

Lieferant: Abnova
ABNOMAB5157EA 1030 EUR
ABNOMAB5157
Anti-ABCC1 Monoclonal Antibody [clone: QCRL-3]
Antikörper
Mouse monoclonal antibody raised against recombinant ABCC1.

Recommended Dilutions: Flow Cytometry: 1:20-1:50

Type: Primary
Antigen: ABCC1
Clonality: Monoclonal
Clone: QCRL-3
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR