Anti-ABCC6 Rat Monoclonal Antibody [clone: M6II-31]

Lieferant: Abnova
ABNOMAB3476EA 1020 EUR
ABNOMAB3476
Anti-ABCC6 Rat Monoclonal Antibody [clone: M6II-31]
Antikörper
Rat monoclonal antibody raised against recombinant ABCC6.

Recommended Dilutions: Immunocytochemistry: 1:20-50; Western Blot: 1:20-50; Immunohistochemistry (Frozen sections): 1:20-50; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:20

Type: Primary
Antigen: ABCC6
Clonality: Monoclonal
Clone: M6II-31
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR