Anti-ABCC3 Mouse Monoclonal Antibody [clone: M3II-9]

Lieferant: Abnova
ABNOMAB3384EA 684 EUR
ABNOMAB3384
Anti-ABCC3 Mouse Monoclonal Antibody [clone: M3II-9]
Antikörper
Mouse monoclonal antibody raised against partial recombinant ABCC3.

Recommended Dilutions: Immunocytochemistry: 1:20-1:50; Immunohistochemistry (Frozen sections): 1:20; Western Blot: 1:20-1:50

Type: Primary
Antigen: ABCC3
Clonality: Monoclonal
Clone: M3II-9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR