Anti-ABCC2 Mouse Monoclonal Antibody [clone: M2II-12]

Lieferant: Abnova
ABNOMAB3383EA 684 EUR
ABNOMAB3383
Anti-ABCC2 Mouse Monoclonal Antibody [clone: M2II-12]
Antikörper
Mouse monoclonal antibody raised against partial recombinant ABCC2.

Recommended Dilutions: Western Blot: 1:20-50; Immunohistochemistry (Frozen sections): 1:20; Immunocytochemistry: 1:20-1:50

Type: Primary
Antigen: ABCC2
Clonality: Monoclonal
Clone: M2II-12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR