Anti-BKPyVgp4 Mouse Monoclonal Antibody [clone: 6E10]

Lieferant: Abnova
ABNOMAB3204-M22EA 382 EUR
ABNOMAB3204-M22
Anti-BKPyVgp4 Mouse Monoclonal Antibody [clone: 6E10]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant BKV VP1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: BKPyVgp4
Clonality: Monoclonal
Clone: 6E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR