Anti-ACTB Mouse Monoclonal Antibody [clone: 8H10D10]

Lieferant: Abnova
ABNOMAB10501EA 370 EUR
ABNOMAB10501
Anti-ACTB Mouse Monoclonal Antibody [clone: 8H10D10]
Antikörper
Mouse monoclonal antibody raised against synthetic peptide of ACTB.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400

Type: Primary
Antigen: ACTB
Clonality: Monoclonal
Clone: 8H10D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Hamster, Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR