Anti-BCL2 Mouse Monoclonal Antibody [clone: 8E12]

Lieferant: Abnova
ABNOMAB10499EA 370 EUR
ABNOMAB10499
Anti-BCL2 Mouse Monoclonal Antibody [clone: 8E12]
Antikörper
Mouse monoclonal antibody raised against synthetic peptide of BCL2.

Recommended Dilutions: ELISA: 1:10000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400

Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: 8E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR