Anti-ATP2C1 Mouse Monoclonal Antibody [clone: 4G12]
Lieferant: Abnova
ABNOMAB10496EA 518 EUR
ABNOMAB10496
Anti-ATP2C1 Mouse Monoclonal Antibody [clone: 4G12]
Antikörper
Mouse monoclonal antibody raised against partial recombinant ATP2C1.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000
Type: Primary
Antigen: ATP2C1
Clonality: Monoclonal
Clone: 4G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey